Gene Transcripts
- Gene Symbol
- Gprin2
- Entrez Gene ID
- 432839
- Gene Name
- G protein regulated inducer of neurite outgrowth 2
- Gene Aliases
- C130040D06Rik, C230073P13, Gm286, mKIAA0514
- Chromosome Location
- Chr. 14: 34185688 - 34201692 on Build GRCm38
- UniGene ID
- Mm.294643
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK048200.1 |
- |
1 |
1084 |
|
AK082455.1 |
- |
1 |
347 |
|
BC066049.1 |
- |
2 |
2732 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CX730911.1 |
- |
1 |
451 |
|
Chr. 14 - 34186136 - 34186770 [-] |
GenBankmRNA |
BB651025.1 |
- |
1 |
345 |
|
Chr. 14 - 34185987 - 34186664 [-] |
GenBankmRNA |
BB367967.2 |
- |
1 |
433 |
|
Chr. 14 - 34186104 - 34186741 [-] |
View Full Product Details