Gene Transcripts
- Gene Symbol
- Rspo2
- Entrez Gene ID
- 239405
- Gene Name
- R-spondin 2 homolog (Xenopus laevis)
- Gene Aliases
- 2610028F08Rik, AA673245, D430027K22, ftls
- Chromosome Location
- Chr. 15: 43020788 - 43171595 on Build GRCm38
- UniGene ID
- Mm.193274
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011587.2 |
- |
1 |
849 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY754718.1 |
- |
1 |
443 |
|
Chr. 15 - 43170534 - 43171224 [+] |
View Full Product Details