Gene Transcripts
- Gene Symbol
- Senp1
- Entrez Gene ID
- 223870
- Gene Name
- SUMO1/sentrin specific peptidase 1
- Gene Aliases
- 2310046A20Rik, D15Ertd528e, E330036L07Rik, suPr-2
- Chromosome Location
- Chr. 15: 98038744 - 98093569 on Build GRCm38
- UniGene ID
- Mm.384023
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009838.1 |
- |
1 |
491 |
|
View Full Product Details