Gene Transcripts

Gene Symbol
Zfp53
Entrez Gene ID
24132
Gene Name
zinc finger protein 53
Gene Aliases
D030067O06Rik, KRAZ1, Zfp-53, Zfp118, zfas8
Chromosome Location
Chr. 17: 21488988 - 21510477 on Build GRCm38
UniGene ID
Mm.42140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090292.1 - 919
BC023798.1 - 817


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CF908887.1 - 6 671 Chr. 17 - 21489119 - 21530431 [+]

View Full Product Details