Gene Transcripts
- Gene Symbol
- H60c
- Entrez Gene ID
- 670558
- Gene Name
- histocompatibility 60c
- Gene Aliases
- 4632413I24Rik, Gm9491
- Chromosome Location
- Chr. 10: 3256202 - 3268092 on Build GRCm38
- UniGene ID
- Mm.398582
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014573.1 |
- |
3 |
1514 |
|
View Full Product Details