Gene Transcripts
- Gene Symbol
- Nkx1-2
- Entrez Gene ID
- 20231
- Gene Name
- NK1 transcription factor related, locus 2 (Drosophila)
- Gene Aliases
- Nkx-1.1, Nkx-1.2, Sax1
- Chromosome Location
- Chr. 7: 132596239 - 132599637 on Build GRCm38
- UniGene ID
- Mm.334281
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF222442.1 |
- |
|
1219 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV308505.2 |
- |
1 |
250 |
|
Chr. 7 - 132594882 - 132595624 [-] |
View Full Product Details