Gene Transcripts
- Gene Symbol
- Rabepk
- Entrez Gene ID
- 227746
- Gene Name
- Rab9 effector protein with kelch motifs
- Gene Aliases
- 8430412M01Rik, 9530020D24Rik, AV073337, C87311, Rab9p40
- Chromosome Location
- Chr. 2: 34777892 - 34799912 on Build GRCm38
- UniGene ID
- Mm.158705
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018405.1 |
- |
1 |
226 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY720528.1 |
- |
1 |
221 |
|
Chr. 2 - 34785801 - 34786550 [-] |
View Full Product Details