Gene Transcripts
- Gene Symbol
- Mgat4c
- Entrez Gene ID
- 67569
- Gene Name
- MGAT4 family, member C
- Gene Aliases
- 9130411I17Rik, GntIVh
- Chromosome Location
- Chr. 10: 101681487 - 102391851 on Build GRCm38
- UniGene ID
- Mm.25252
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080684.1 |
- |
1 |
38 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY725284.1 |
- |
1 |
35 |
|
Chr. 10 - 101681489 - 102011876 [+] |
View Full Product Details