Gene Transcripts
- Gene Symbol
- Shkbp1
- Entrez Gene ID
- 192192
- Gene Name
- Sh3kbp1 binding protein 1
- Gene Aliases
- B930062H15Rik, Sb1
- Chromosome Location
- Chr. 7: 27342128 - 27356015 on Build GRCm38
- UniGene ID
- Mm.21297
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK048719.1 |
- |
2 |
190 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB650638.1 |
- |
2 |
190 |
|
Chr. 7 - 27354350 - 27355848 [+] |
View Full Product Details