Gene Transcripts

Gene Symbol
Hotair
Entrez Gene ID
100503872
Gene Name
HOX transcript antisense RNA (non-protein coding)
Gene Aliases
Gm16258, OTTMUSG00000032781
Chromosome Location
Chr. 15: 102944062 - 102947946 on Build GRCm38
UniGene ID
Mm.383980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_047528.1 1, 2 459
GenBank mRNA AK035706.1 - 1, 2 243
BB628325.1 - 1, 2 240

View Full Product Details