Gene Transcripts
- Gene Symbol
- Clgn
- Entrez Gene ID
- 12745
- Gene Name
- calmegin
- Gene Aliases
- 4930459O04Rik, AI528775, Cln
- Chromosome Location
- Chr. 8: 83389826 - 83428552 on Build GRCm38
- UniGene ID
- Mm.358581
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015491.1 |
- |
1 |
608 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715039.1 |
- |
1 |
606 |
|
Chr. 8 - 83427552 - 83428343 [+] |
View Full Product Details