Gene Transcripts
- Gene Symbol
- Irf2bp2
- Entrez Gene ID
- 270110
- Gene Name
- interferon regulatory factor 2 binding protein 2
- Gene Aliases
- E130305N23Rik, IRF-2BP2
- Chromosome Location
- Chr. 8: 126588296 - 126593436 on Build GRCm38
- UniGene ID
- Mm.334918
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK050834.1 |
- |
|
716 |
|
View Full Product Details