Gene Transcripts

Gene Symbol
Tln1
Entrez Gene ID
21894
Gene Name
talin 1
Gene Aliases
Tln
Chromosome Location
Chr. 4: 43531513 - 43562583 on Build GRCm38
UniGene ID
Mm.208601
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK006088.1 - 2 538
BC055348.1 - 2 333
BC059711.1 - 2 198


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU559994.1 - 2 304 Chr. 4 - 43559355 - 43562348 [-]
GenBankmRNA W83462.1 - 194 Chr. - - [-]
GenBankmRNA BY251028.1 - 2 205 Chr. 4 - 43559417 - 43562250 [-]
GenBankmRNA BI648366.1 - 2 331 Chr. 4 - 43559343 - 43562378 [-]
GenBankmRNA CF739684.1 - 2 201 Chr. 4 - 43559352 - 43562247 [-]
GenBankmRNA BY039994.1 - 2 148 Chr. 4 - 43559365 - 43562193 [-]
GenBankmRNA BY274827.1 - 2 147 Chr. 4 - 43559360 - 43562193 [-]
GenBankmRNA CK793097.1 - 2 349 Chr. 4 - 43559350 - 43562395 [-]
GenBankmRNA BY246090.1 - 2 212 Chr. 4 - 43559414 - 43562257 [-]
GenBankmRNA CB274742.1 - 2 173 Chr. 4 - 43559509 - 43562219 [-]
GenBankmRNA CB274594.1 - 2 173 Chr. 4 - 43559509 - 43562219 [-]
GenBankmRNA BU961458.1 - 2 201 Chr. 4 - 43559350 - 43562244 [-]

View Full Product Details