Gene Transcripts
- Gene Symbol
- Zfp53
- Entrez Gene ID
- 24132
- Gene Name
- zinc finger protein 53
- Gene Aliases
- D030067O06Rik, KRAZ1, Zfp-53, Zfp118, zfas8
- Chromosome Location
- Chr. 17: 21488988 - 21510477 on Build GRCm38
- UniGene ID
- Mm.42140
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK083692.1 |
- |
|
2043 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB448675.2 |
- |
1 |
509 |
|
Chr. 17 - 21532557 - 21533184 [+] |
View Full Product Details