Gene Transcripts
- Gene Symbol
- Eif3a
- Entrez Gene ID
- 13669
- Gene Name
- eukaryotic translation initiation factor 3, subunit A
- Gene Aliases
- A830012B05Rik, Csma, Eif3, Eif3s10, mKIAA0139
- Chromosome Location
- Chr. 19: 60761116 - 60790693 on Build GRCm38
- UniGene ID
- Mm.2238
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043609.1 |
- |
1 |
971 |
|
AK087340.1 |
- |
1 |
938 |
|
View Full Product Details