Gene Transcripts
- Gene Symbol
- Ipmk
- Entrez Gene ID
- 69718
- Gene Name
- inositol polyphosphate multikinase
- Gene Aliases
- 2410017C19Rik, AA408208, Impk
- Chromosome Location
- Chr. 10: 71347765 - 71385885 on Build GRCm38
- UniGene ID
- Mm.245867
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076771.1 |
- |
|
411 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV264160.2 |
- |
3 |
436 |
|
Chr. 10 - 71381749 - 71433180 [+] |
View Full Product Details