Gene Transcripts
- Gene Symbol
- Adamts5
- Entrez Gene ID
- 23794
- Gene Name
- a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2)
- Gene Aliases
- 9530092O11Rik, ADAM-TS5, ADAMTS1, ADAMTS11, AI481094, ASMP-2
- Chromosome Location
- Chr. 16: 85858157 - 85902374 on Build GRCm38
- UniGene ID
- Mm.112933
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031656.1 |
- |
|
1885 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI732482.1 |
- |
1 |
94 |
|
Chr. 16 - 85902539 - 85903074 [+] |
View Full Product Details