Gene Transcripts
- Gene Symbol
- Cdc73
- Entrez Gene ID
- 214498
- Gene Name
- cell division cycle 73, Paf1/RNA polymerase II complex component
- Gene Aliases
- 8430414L16Rik, BC027756, C130030P16Rik, C81219, Hrpt2
- Chromosome Location
- Chr. 1: 143607499 - 143702888 on Build GRCm38
- UniGene ID
- Mm.156727
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018410.1 |
- |
|
583 |
|
View Full Product Details