Gene Transcripts
- Gene Symbol
- Hrct1
- Entrez Gene ID
- 100039781
- Gene Name
- histidine rich carboxyl terminus 1
- Gene Aliases
- 3830408D24Rik
- Chromosome Location
- Chr. 4: 43727198 - 43728110 on Build GRCm38
- UniGene ID
- Mm.18796
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014425.1 |
- |
|
1352 |
|
View Full Product Details