Gene Transcripts
- Gene Symbol
- Lamtor3
- Entrez Gene ID
- 56692
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
- Gene Aliases
- AW556229, Map2k1ip1, Mapbp, Mapksp1, Mp1
- Chromosome Location
- Chr. 3: 137918555 - 137928762 on Build GRCm38
- UniGene ID
- Mm.331392
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031303.1 |
- |
1 |
934 |
|
View Full Product Details