Gene Transcripts
- Gene Symbol
- Atf7
- Entrez Gene ID
- 223922
- Gene Name
- activating transcription factor 7
- Gene Aliases
- 1110012F10Rik, 9430065F09Rik, AI549878, C130020M04Rik
- Chromosome Location
- Chr. 15: 102536643 - 102625421 on Build GRCm38
- UniGene ID
- Mm.208714
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003630.1 |
- |
|
18 |
|
AK154588.1 |
- |
|
5164 |
|
AK170250.1 |
- |
|
110 |
|
View Full Product Details