Gene Transcripts
- Gene Symbol
- 5830453K13Rik
- Entrez Gene ID
- 76076
- Gene Name
- RIKEN cDNA 5830453K13 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018007.1 |
- |
1 |
274 |
|
- Gene Symbol
- Cdc26
- Entrez Gene ID
- 66440
- Gene Name
- cell division cycle 26
- Gene Aliases
- 2010012C09Rik
- Chromosome Location
- Chr. 4: 62393812 - 62408640 on Build GRCm38
- UniGene ID
- Mm.109930
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041840.1 |
- |
2 |
2672 |
|
View Full Product Details