Gene Transcripts
- Gene Symbol
- Snapc3
- Entrez Gene ID
- 77634
- Gene Name
- small nuclear RNA activating complex, polypeptide 3
- Gene Aliases
- 1810020H02Rik, 4930558A07Rik, 5031401C21Rik, AI414457, E030018J20Rik
- Chromosome Location
- Chr. 4: 83417701 - 83467658 on Build GRCm38
- UniGene ID
- Mm.271985
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK048571.1 |
- |
1 |
1566 |
|
AK053153.1 |
- |
1 |
2249 |
|
AK171985.1 |
- |
1 |
1617 |
|
View Full Product Details