Gene Transcripts
- Gene Symbol
- Foxp1
- Entrez Gene ID
- 108655
- Gene Name
- forkhead box P1
- Gene Aliases
- 3110052D19Rik, 4932443N09Rik, AI461938, AW494214
- Chromosome Location
- Chr. 6: 98925342 - 99523024 on Build GRCm38
- UniGene ID
- Mm.234965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046104.1 |
- |
1 |
958 |
|
AK052184.1 |
- |
1 |
1463 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB474071.2 |
- |
1 |
24 |
|
Chr. 6 - 98925352 - 98925980 [-] |
View Full Product Details