Gene Transcripts
- Gene Symbol
- Coro2a
- Entrez Gene ID
- 107684
- Gene Name
- coronin, actin binding protein 2A
- Gene Aliases
- 9030208C03Rik, AI563590, IR10
- Chromosome Location
- Chr. 4: 46536937 - 46601929 on Build GRCm38
- UniGene ID
- Mm.416311
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045728.1 |
- |
2 |
4526 |
|
View Full Product Details