Gene Transcripts
- Gene Symbol
- STARD7-AS1
- Entrez Gene ID
- 285033
- Gene Name
- STARD7 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 96208416 - 96242623 on Build GRCh38
- UniGene ID
- Hs.58648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046322.1 |
|
4 |
2292 |
|
GenBank mRNA |
AK057419.1 |
- |
4 |
1764 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000432267.2 |
- |
2 |
1533 |
|
Chr. 2 - 96239713 - 96242621 [+] |
GENCODE |
ENST00000446816.1 |
- |
3 |
1366 |
|
Chr. 2 - 96208416 - 96242594 [+] |
View Full Product Details