Gene Transcripts
- Gene Symbol
- SFTA1P
- Entrez Gene ID
- 207107
- Gene Name
- surfactant associated 1, pseudogene
- Gene Aliases
- SFTPF
- Chromosome Location
- Chr. 10: 10784439 - 10794914 on Build GRCh38
- UniGene ID
- Hs.31562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027082.1 |
|
2 |
348 |
|
GenBank mRNA |
AY102069.1 |
- |
2 |
348 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000446372.2 |
- |
2 |
348 |
|
Chr. 10 - 10784439 - 10794914 [-] |
View Full Product Details