Gene Transcripts
- Gene Symbol
- LINC01554
- Entrez Gene ID
- 202299
- Gene Name
- long intergenic non-protein coding RNA 1554
- Gene Aliases
- C5orf27, FIS
- Chromosome Location
- Chr. 5: 95852232 - 95860133 on Build GRCh38
- UniGene ID
- Hs.8373
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000620167.1 |
- |
|
131 |
|
Chr. - - [-] |
GENCODE |
ENST00000357880.7 |
- |
|
605 |
|
Chr. 16 - 58749709 - 58750122 [+] |
View Full Product Details