Gene Transcripts

Gene Symbol
C9orf62
Entrez Gene ID
157927
Gene Name
chromosome 9 open reading frame 62
Gene Aliases
Chromosome Location
Chr. 9: 135343249 - 135346562 on Build GRCh38
UniGene ID
Hs.559511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173520.2 NP_775791.1 2 463
GenBank mRNA BC034752.1 - 2 463


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000623103.1 - 463 Chr. - - [-]

View Full Product Details