Gene Transcripts
- Gene Symbol
- C10orf91
- Entrez Gene ID
- 170393
- Gene Name
- chromosome 10 open reading frame 91
- Gene Aliases
- bA432J24.4
- Chromosome Location
- Chr. 10: 132445210 - 132448321 on Build GRCh38
- UniGene ID
- Hs.375059
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030794.1 |
- |
3 |
842 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000321248.4 |
- |
3 |
842 |
|
Chr. 10 - 132445189 - 132449386 [+] |
View Full Product Details