Gene Transcripts

Gene Symbol
TP53TG1
Entrez Gene ID
11257
Gene Name
TP53 target 1 (non-protein coding)
Gene Aliases
LINC00096, NCRNA00096, P53TG1, P53TG1-D, TP53AP1
Chromosome Location
Chr. 7: 87325347 - 87345492 on Build GRCh38
UniGene ID
Hs.274329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015381.1 3 657
GenBank mRNA AB007455.1 - 3 680
AB007456.1 - 4 609
AB007457.1 - 3 460
AB007458.1 - 3 389
BC002709.2 - 3 657
BC061927.1 - 3 610
BC103631.1 - 3 364


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM912218.1 - 592 Chr. - - [-]
GENCODE ENST00000359941.9 - 680 Chr. 7 - 87341647 - 87345378 [-]
GenBankmRNA BE786049.1 - 3 635 Chr. 7 - 87325318 - 87345473 [-]
GenBankmRNA BM850328.1 - 3 353 Chr. 7 - 87325370 - 87345188 [-]
GENCODE ENST00000432193.5 - 3 460 Chr. 7 - 87325350 - 87345486 [-]
GenBankmRNA BM912196.1 - 592 Chr. - - [-]
GENCODE ENST00000416560.5 - 4 609 Chr. 7 - 87325299 - 87345515 [-]
GenBankmRNA N47733.1 - 3 247 Chr. 7 - 87325237 - 87345082 [-]
GenBankmRNA BM915584.1 - 3 592 Chr. 7 - 87325297 - 87345427 [-]
GENCODE ENST00000421293.1 - 3 389 Chr. 7 - 87325350 - 87345486 [-]
GenBankmRNA CN409085.1 - 3 638 Chr. 7 - 87325365 - 87345473 [-]
GenBankmRNA AA447887.1 - 3 440 Chr. 7 - 87325353 - 87345275 [-]
GenBankmRNA BQ961841.1 - 3 665 Chr. 7 - 87325297 - 87345500 [-]
GenBankmRNA BM550903.1 - 3 592 Chr. 7 - 87325297 - 87345419 [-]
GenBankmRNA BU536344.1 - 3 369 Chr. 7 - 87325347 - 87345201 [-]
GenBankmRNA BM550333.1 - 3 593 Chr. 7 - 87325261 - 87345428 [-]
GenBankmRNA BX110032.1 - 3 624 Chr. 7 - 87325347 - 87345459 [-]

View Full Product Details