Gene Transcripts

Gene Symbol
C2orf27B
Entrez Gene ID
408029
Gene Name
chromosome 2 open reading frame 27B
Gene Aliases
Chromosome Location
Chr. 2: 131794961 - 131801661 on Build GRCh38
UniGene ID
Hs.145567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_214461.2 NP_999626.1 2 343
GenBank mRNA BC043584.1 - 2 343


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000623129.1 - 343 Chr. - - [-]

View Full Product Details