Gene Transcripts

Gene Symbol
MUC2
Entrez Gene ID
4583
Gene Name
mucin 2, oligomeric mucus/gel-forming
Gene Aliases
MLP, MUC-2, SMUC
Chromosome Location
Chr. 11: 1074875 - 1110508 on Build GRCh38
UniGene ID
Hs.315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002457.3 NP_002448.3 13770
GenBank mRNA AF257167.1 - 6 1191
L21998.1 - 13770
M86523.1 - 2 270
M94132.1 - 6 1158


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000616479.1 - 270 Chr. - - [-]

View Full Product Details