Gene Transcripts
- Gene Symbol
- GAS1RR
- Entrez Gene ID
- 100506834
- Gene Name
- GAS1 adjacent regulatory RNA
- Gene Aliases
- LncRNA-Hh
- Chromosome Location
- Chr. 9: 86948698 - 87002033 on Build GRCh38
- UniGene ID
- Hs.553987
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_049794.1 |
|
4 |
711 |
|
GenBank mRNA |
BC036850.1 |
- |
4 |
722 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000415801.1 |
- |
4 |
710 |
|
Chr. 9 - 86948699 - 87002033 [+] |
View Full Product Details