Gene Transcripts
- Gene Symbol
- SMIM2-AS1
- Entrez Gene ID
- 101929212
- Gene Name
- SMIM2 antisense RNA 1
- Gene Aliases
- C13orf44-AS1, C3orf81-AS1
- Chromosome Location
- Chr. 13: 44110549 - 44154079 on Build GRCh38
- UniGene ID
- Hs.534877
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104064.1 |
|
2 |
435 |
|
NR_104065.1 |
|
2 |
130 |
|
GenBank mRNA |
DB036693.1 |
- |
2 |
435 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000444663.5 |
- |
2 |
217 |
|
Chr. 13 - 44142546 - 44148030 [+] |
GenBankmRNA |
AA399284.1 |
- |
2 |
126 |
|
Chr. 13 - 44142637 - 44148028 [+] |
View Full Product Details