Gene Transcripts

Gene Symbol
GUSBP11
Entrez Gene ID
91316
Gene Name
glucuronidase, beta pseudogene 11
Gene Aliases
Chromosome Location
Chr. 22: 23638487 - 23717423 on Build GRCh38
UniGene ID
Hs.148656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024448.2 7 1129
GenBank mRNA AK056875.1 - 7 1062
BC063045.1 - 7 892


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000445682.2 - 7 1062 Chr. 22 - 23638487 - 23717356 [-]
GENCODE ENST00000452737.5 - 7 894 Chr. 22 - 23638487 - 23717321 [-]

View Full Product Details