Gene Transcripts
- Gene Symbol
- HCG22
- Entrez Gene ID
- 285834
- Gene Name
- HLA complex group 22
- Gene Aliases
- PBMUCL2
- Chromosome Location
- Chr. 6: 31053450 - 31059890 on Build GRCh38
- UniGene ID
- Hs.207528
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000426185.1 |
- |
3 |
1890 |
|
Chr. 6 - 31054207 - 31059876 [+] |
GENCODE |
ENST00000565192.1 |
- |
4 |
1571 |
|
Chr. 6 - 31053450 - 31059890 [+] |
View Full Product Details