Gene Transcripts
- Gene Symbol
- EPHA1-AS1
- Entrez Gene ID
- 285965
- Gene Name
- EPHA1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 143407813 - 143523449 on Build GRCh38
- UniGene ID
- Hs.642649
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033897.1 |
|
5 |
3503 |
|
GenBank mRNA |
AL833583.1 |
- |
5 |
3504 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000429289.5 |
- |
5 |
3503 |
|
Chr. 7 - 143407813 - 143523449 [+] |
View Full Product Details