Gene Transcripts
- Gene Symbol
- MYCNOS
- Entrez Gene ID
- 10408
- Gene Name
- MYCN opposite strand
- Gene Aliases
- MYCN-AS1, N-CYM, NCYM, NYCM
- Chromosome Location
- Chr. 2: 15936265 - 15941723 on Build GRCh38
- UniGene ID
- Hs.651453
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX279550.1 |
- |
1 |
239 |
|
Chr. 2 - 15940121 - 15941238 [-] |
GENCODE |
ENST00000419083.5 |
- |
2 |
290 |
|
Chr. 2 - 15939898 - 15941723 [-] |
View Full Product Details