Gene Transcripts
- Gene Symbol
- FRY-AS1
- Entrez Gene ID
- 100507099
- Gene Name
- FRY antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 13: 32024046 - 32031639 on Build GRCh38
- UniGene ID
- Hs.536364
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103839.1 |
|
4 |
427 |
|
GenBank mRNA |
BC035084.1 |
- |
5 |
432 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000428419.1 |
- |
4 |
427 |
|
Chr. 13 - 32025314 - 32031639 [-] |
View Full Product Details