Gene Transcripts

Gene Symbol
KCNQ1DN
Entrez Gene ID
55539
Gene Name
KCNQ1 downstream neighbor (non-protein coding)
Gene Aliases
BWRT, HSA404617
Chromosome Location
Chr. 11: 2870033 - 2872105 on Build GRCh38
UniGene ID
Hs.127821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024627.1 1 216
GenBank mRNA AB039920.1 - 1 216
AJ404617.2 - 1 186
BC098159.1 - 1 78
BC098256.1 - 1 78
BC098319.1 - 1 82
BC098344.1 - 1 78


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000441418.2 - 1 216 Chr. 11 - 2870033 - 2872105 [+]
GenBankmRNA AI791256.1 - 1 216 Chr. 11 - 2870033 - 2870481 [+]

View Full Product Details