Gene Transcripts
- Gene Symbol
- MYLK-AS1
- Entrez Gene ID
- 100506826
- Gene Name
- MYLK antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 123585513 - 123630821 on Build GRCh38
- UniGene ID
- Hs.556600
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ186840.1 |
- |
3 |
526 |
|
Chr. 3 - 123585584 - 123630777 [+] |
GenBankmRNA |
BI599102.1 |
- |
3 |
558 |
|
Chr. 3 - 123585554 - 123630820 [+] |
GENCODE |
ENST00000470449.1 |
- |
3 |
554 |
|
Chr. 3 - 123585556 - 123630821 [+] |
View Full Product Details