Gene Transcripts
- Gene Symbol
- IGSF11-AS1
- Entrez Gene ID
- 100506765
- Gene Name
- IGSF11 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 118943076 - 118948241 on Build GRCh38
- UniGene ID
- Hs.660823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046230.1 |
|
4 |
609 |
|
GenBank mRNA |
BC042737.1 |
- |
4 |
631 |
|
BI829854.1 |
- |
4 |
609 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000477009.2 |
- |
4 |
612 |
|
Chr. 3 - 118943073 - 118948241 [+] |
GenBankmRNA |
BG772164.1 |
- |
4 |
636 |
|
Chr. 3 - 118943075 - 118947415 [+] |
View Full Product Details