Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
79863
Gene Name
ribosome binding factor A (putative)
Gene Aliases
C18orf22, HsT169
Chromosome Location
Chr. 18: 80034346 - 80050652 on Build GRCh38
UniGene ID
Hs.728931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171967.1 NP_001165438.1 6 1859
NM_024805.2 NP_079081.2 7 1944


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000562391.5 - 1 18 Chr. 18 - 80046900 - 80081293 [+]

View Full Product Details