Gene Transcripts
- Gene Symbol
- MRPL4
- Entrez Gene ID
- 51073
- Gene Name
- mitochondrial ribosomal protein L4
- Gene Aliases
- CGI-28, L4mt
- Chromosome Location
- Chr. 19: 10251964 - 10260060 on Build GRCh38
- UniGene ID
- Hs.279652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI092267.1 |
- |
1 |
75 |
|
AL561917.2 |
- |
1 |
18 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000587088.1 |
- |
1 |
38 |
|
Chr. 19 - 10259109 - 10260045 [-] |
View Full Product Details