Gene Transcripts
- Gene Symbol
- CIRBP-AS1
- Entrez Gene ID
- 148046
- Gene Name
- CIRBP antisense RNA 1
- Gene Aliases
- C19orf23
- Chromosome Location
- Chr. 19: 1267471 - 1270260 on Build GRCh38
- UniGene ID
- Hs.438829
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027271.1 |
|
1 |
179 |
|
GenBank mRNA |
BC026041.1 |
- |
1 |
178 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000585832.2 |
- |
1 |
160 |
|
Chr. 19 - 1268166 - 1270241 [-] |
View Full Product Details