Gene Transcripts
- Gene Symbol
- LINC00839
- Entrez Gene ID
- 84856
- Gene Name
- long intergenic non-protein coding RNA 839
- Gene Aliases
- Chromosome Location
- Chr. 10: 42475491 - 42495337 on Build GRCh38
- UniGene ID
- Hs.55977
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024638.1 |
- |
1 |
919 |
|
- Gene Symbol
- XYLB
- Entrez Gene ID
- 9942
- Gene Name
- xylulokinase
- Gene Aliases
- Chromosome Location
- Chr. 3: 38346760 - 38453041 on Build GRCh38
- UniGene ID
- Hs.659985
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000594379.1 |
- |
7 |
748 |
|
Chr. 19 - 52300693 - 52345229 [-] |
View Full Product Details