Gene Transcripts
- Gene Symbol
- Fndc3a
- Entrez Gene ID
- 319448
- Gene Name
- fibronectin type III domain containing 3A
- Gene Aliases
- 1700094E19Rik, D14Ertd453e, F730017H24Rik, Fndc3, sys
- Chromosome Location
- Chr. 14: 72537948 - 72710686 on Build GRCm38
- UniGene ID
- Mm.205421
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007062.1 |
- |
2 |
374 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM875133.1 |
- |
1 |
235 |
|
Chr. 14 - 72603024 - 72603377 [+] |
View Full Product Details