Gene Transcripts
- Gene Symbol
- V1ra8
- Entrez Gene ID
- 113850
- Gene Name
- vomeronasal 1 receptor, A8
- Gene Aliases
- Vmn1r-ps33
- Chromosome Location
- Chr. 6: 90202817 - 90203656 on Build GRCm38
- UniGene ID
- Mm.377146
- Species
- Mus musculus
- Gene Symbol
- Vmn1r52
- Entrez Gene ID
- 113849
- Gene Name
- vomeronasal 1 receptor 52
- Gene Aliases
- V1ra7, VN3
- Chromosome Location
- Chr. 6: 90178716 - 90179645 on Build GRCm38
- UniGene ID
- Mm.435482
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053222.1 |
NP_444452.1
|
1 |
205 |
|
View Full Product Details