Gene Transcripts
- Gene Symbol
- Vmn1r41
- Entrez Gene ID
- 113857
- Gene Name
- vomeronasal 1 receptor 41
- Gene Aliases
- V1ra12, V1rb9
- Chromosome Location
- Chr. 6: 89740888 - 89747414 on Build GRCm38
- UniGene ID
- Mm.377154
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053230.2 |
NP_444460.2
|
2 |
546 |
|
View Full Product Details